Gene Transcripts

Gene Symbol
Ikzf4
Entrez Gene ID
22781
Gene Name
IKAROS family zinc finger 4
Gene Aliases
A630026H08Rik, Eos, Zfpn1a4, Znfn1a4
Chromosome Location
Chr. 10: 128630839 - 128669358 on Build GRCm38
UniGene ID
Mm.358648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011772.2 NP_035902.2 2, 3 360
GenBank mRNA AB017615.1 - 1, 2 767
AK089522.1 - 2, 3 194
AK165740.1 - 624
BC030070.1 - 1 558
BC141286.1 - 1, 2 458 9055898
BC145614.1 - 1, 2 458

View Full Product Details