Gene Transcripts

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
Chromosome Location
Chr. 11: 17734810 - 17783055 on Build GRCh38
UniGene ID
Hs.552896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112741.1 NP_001106212.1 2 1204
NM_004976.4 NP_004967.1 2 1204
XM_011520080.2 XP_011518382.1 2 2342
GenBank mRNA BC107129.2 - 2 1221
L00621.1 - 2 1217
M96747.1 - 1 420
S56770.1 - 2 1217

View Full Product Details