Gene Transcripts

Gene Symbol
Psg18
Entrez Gene ID
26438
Gene Name
pregnancy specific glycoprotein 18
Gene Aliases
Cea-3, Cea3, mmCGM6
Chromosome Location
Chr. 7: 18345744 - 18355038 on Build GRCm38
UniGene ID
Mm.89991
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011963.2 NP_036093.2 2 488
GenBank mRNA AF128236.1 - 2 356
BC138857.1 - 2 422 8861879
BY713630.1 - 2 490
CN704107.1 - 2 474
M83346.1 - 2 356

View Full Product Details