Gene Transcripts

Gene Symbol
Lsm2
Entrez Gene ID
27756
Gene Name
LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
D17H6S56E-2, D17H6S56E2, Dmapl, Dmpkap, G7b, Sm-X5, SmX5, snRNP
Chromosome Location
Chr. 17: 34981854 - 34985891 on Build GRCm38
UniGene ID
Mm.165735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110101.2 NP_001103571.1 3, 4 336
NM_001204273.1 NP_001191202.1 4, 5 371
NM_001204274.1 NP_001191203.1 3, 4 336
NM_030597.3 NP_085100.1 3, 4 342
GenBank mRNA AF204146.1 - 3, 4 313
AF204147.1 - 3, 4 313
AF204148.1 - 3, 4 313
AF204149.1 - 3, 4 313
AF204150.1 - 3, 4 313
AF204151.1 - 3, 4 313
AF204152.1 - 3, 4 313
AF204153.1 - 2, 3 555
AF204154.1 - 4, 5 342
AF204155.1 - 4, 5 342
AF204157.1 - 3, 4 180
AK151605.1 - 3, 4 310
AK178855.1 - 3, 4 124
BB848823.1 - 3, 4 342
BC014288.1 - 3, 4 321 3982410
BC049543.1 - 335 6774564
BY079930.1 - 2, 3 270

View Full Product Details