Gene Transcripts

Gene Symbol
Timm13
Entrez Gene ID
30055
Gene Name
translocase of inner mitochondrial membrane 13
Gene Aliases
D10Ertd378e, Tim9, Timm9
Chromosome Location
Chr. 10: 80899450 - 80900969 on Build GRCm38
UniGene ID
Mm.142132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013895.4 NP_038923.1 1, 2 354
GenBank mRNA AF144702.1 - 1, 2 356
AK003054.1 - 1, 2 160
AK009041.1 - 1, 2 148
AK009152.1 - 1, 2 164
AK013230.1 - 1, 2 152
AK088984.1 - 1, 2 164
BC011436.1 - 1, 2 167 2812494

View Full Product Details