Gene Transcripts
- Gene Symbol
- KIF3C
- Entrez Gene ID
- 3797
- Gene Name
- kinesin family member 3C
- Gene Aliases
- Chromosome Location
- Chr. 2: 25926586 - 25982574 on Build GRCh38
- UniGene ID
- Hs.21611
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002254.6 |
NP_002245.4
|
6, 7 |
2774 |
|
XM_005264299.2 |
XP_005264356.2
|
6, 7 |
2771 |
|
GenBank mRNA |
AF018164.1 |
- |
6, 7 |
2268 |
|
AF035621.1 |
- |
6, 7 |
2270 |
|
AK293738.1 |
- |
7, 8 |
2038 |
|
AK315324.1 |
- |
7, 8 |
2798 |
|
BC018932.2 |
- |
7, 8 |
750 |
|
BC042486.1 |
- |
7, 8 |
2681 |
3866587 |
BC092406.1 |
- |
6, 7 |
2659 |
6504352 |
BC130423.1 |
- |
6, 7 |
2345 |
40146454 |
BC132783.1 |
- |
6, 7 |
2345 |
40146805 |
BC144223.1 |
- |
6, 7 |
2339 |
9052743 |
BX571741.1 |
- |
6, 7 |
2697 |
|
View Full Product Details