Gene Transcripts

Gene Symbol
KLRB1
Entrez Gene ID
3820
Gene Name
killer cell lectin like receptor B1
Gene Aliases
CD161, CLEC5B, NKR, NKR-P1, NKR-P1A, NKRP1A, hNKR-P1A
Chromosome Location
Chr. 12: 9595274 - 9607901 on Build GRCh38
UniGene ID
Hs.169824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002258.2 NP_002249.1 6 667
GenBank mRNA AK292022.1 - 6 682
BC113997.1 - 6 655
BC114516.1 - 6 655 40031215
U11276.1 - 6 665

View Full Product Details