Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001293683.1 | NP_001280612.1 | 7 | 756 | |
NM_001293684.1 | NP_001280613.1 | 7 | 756 | ||
NM_025635.4 | NP_079911.1 | 7 | 756 | ||
GenBank mRNA | AK004305.1 | - | 7 | 708 | |
AK008144.1 | - | 7 | 706 | ||
AK013556.1 | - | 7 | 691 | ||
AK075977.1 | - | 7 | 702 | ||
AK082215.1 | - | 7 | 705 | ||
AK134380.1 | - | 7 | 708 | ||
AK170637.1 | - | 3 | 248 | ||
BC013559.1 | - | 7 | 674 | ||
BC027100.1 | - | 7 | 713 | 5359154 | |
BC034870.1 | - | 7 | 685 | 1382192 |