Gene Transcripts

Gene Symbol
Pla2g6
Entrez Gene ID
53357
Gene Name
phospholipase A2, group VI
Gene Aliases
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta
Chromosome Location
Chr. 15: 79286228 - 79328530 on Build GRCm38
UniGene ID
Mm.155620
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199023.1 NP_001185952.1 11 2258
NM_001199024.1 NP_001185953.1 10 2093
NM_001199025.1 NP_001185954.1 11 1598
NM_016915.4 NP_058611.1 10 1639
GenBank mRNA AF259401.1 - 10 1538
AK144495.1 - 11 1472
AK153333.1 - 10 1899
BC003487.1 - 10 1432 2811357
BC049778.1 - 10 1819
BC052845.1 - 10 1464 30054573
BC057209.1 - 11 1851 5329147
U88624.2 - 9 1315

View Full Product Details