Gene Transcripts

Gene Symbol
Acin1
Entrez Gene ID
56215
Gene Name
apoptotic chromatin condensation inducer 1
Gene Aliases
2610036I19Rik, 2610510L13Rik, Acinus, Acn, C79325, acinusL, acinusS, mKIAA0670
Chromosome Location
Chr. 14: 54642161 - 54686931 on Build GRCm38
UniGene ID
Mm.297078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085472.2 NP_001078941.2 13 1545
NM_001085473.2 NP_001078942.2 19 3870
NM_001242605.1 NP_001229534.1 12 1609
NM_001242606.1 NP_001229535.1 12 1570
NM_019567.3 NP_062513.3 13 1674
NM_023190.3 NP_075679.2 19 3906
GenBank mRNA AK135284.1 - 2 160
BC094217.1 - 12 1552

View Full Product Details