Gene Transcripts
- Gene Symbol
- Amotl2
- Entrez Gene ID
- 56332
- Gene Name
- angiomotin-like 2
- Gene Aliases
- AW549739, C79691, Lccp, MASCOT, mKIAA0989
- Chromosome Location
- Chr. 9: 102717294 - 102733418 on Build GRCm38
- UniGene ID
- Mm.21145
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019764.2 |
NP_062738.2
|
9, 10 |
2431 |
|
GenBank mRNA |
AF175967.1 |
- |
9, 10 |
1714 |
|
AF175968.1 |
- |
7, 8 |
2571 |
|
AK040912.1 |
- |
9, 10 |
2512 |
|
AK077535.1 |
- |
9, 10 |
2427 |
|
AK141348.1 |
- |
9, 10 |
2431 |
|
AK164279.1 |
- |
9, 10 |
2500 |
|
AK164819.1 |
- |
10, 11 |
2531 |
|
AK169419.1 |
- |
9, 10 |
2431 |
|
AX763686.1 |
- |
8, 9 |
2270 |
|
BC027824.1 |
- |
9, 10 |
2401 |
5323888 |
BC035201.1 |
- |
9, 10 |
2413 |
|
BC108411.1 |
- |
9, 10 |
2465 |
|
View Full Product Details