Gene Transcripts

Gene Symbol
Hsd17b12
Entrez Gene ID
56348
Gene Name
hydroxysteroid (17-beta) dehydrogenase 12
Gene Aliases
2610510O05Rik, AI172963, KIK-I, Kik1
Chromosome Location
Chr. 2: 94032697 - 94157909 on Build GRCm38
UniGene ID
Mm.22505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019657.4 NP_062631.1 8 631
GenBank mRNA AF064635.1 - 8 610
AK012103.1 - 1 45
AK078841.1 - 8 632
AK081869.1 - 8 625
AK082715.1 - 8 625
AK088521.1 - 8 625
AK150849.1 - 8 626
AK152489.1 - 8 626
AK166027.1 - 8 625
AK167908.1 - 8 625
AK188070.1 - 3 108
BC010749.1 - 8 606
BC037620.1 - 7 548 5375831
BC090659.1 - 8 601 30675153

View Full Product Details