Gene Transcripts

Gene Symbol
Retn
Entrez Gene ID
57264
Gene Name
resistin
Gene Aliases
ADSF, Fizz3, Rstn, Xcp4
Chromosome Location
Chr. 8: 3655770 - 3659818 on Build GRCm38
UniGene ID
Mm.1181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204959.1 NP_001191888.1 2, 3 211
NM_022984.4 NP_075360.1 2, 3 211
GenBank mRNA AF290870.1 - 2, 3 204
AF323080.1 - 2, 3 210
AK131613.1 - 2, 3 212
AK131869.1 - 2, 3 208
BC051196.1 - 2, 3 197 1448672

View Full Product Details