Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163233.1 | NP_001156705.1 | 3 | 485 | |
NM_053081.2 | NP_444311.1 | 3 | 485 | ||
GenBank mRNA | AF112439.1 | - | 3 | 294 | |
AY049715.1 | - | 3 | 285 | ||
BC050890.1 | - | 4 | 589 | ||
BC139763.1 | - | 3 | 288 | 40110334 | |
BC141282.1 | - | 3 | 301 | 9055894 |