Gene Transcripts

Gene Symbol
Tnmd
Entrez Gene ID
64103
Gene Name
tenomodulin
Gene Aliases
1110017I01Rik, Bricd4, ChM1L, TeM
Chromosome Location
Chr. X: 133851008 - 133865577 on Build GRCm38
UniGene ID
Mm.46221
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022322.2 NP_071717.1 4 539
GenBank mRNA AB055422.1 - 4 346
AF191768.1 - 4 485
AF219993.1 - 4 412
AF291655.1 - 4 564
AK003748.1 - 4 425
AK014761.1 - 4 378
BC006919.1 - 4 348 3601223
BC049944.1 - 4 539 2811628

View Full Product Details