Gene Transcripts

Gene Symbol
Tma7
Entrez Gene ID
66167
Gene Name
translational machinery associated 7 homolog (S. cerevisiae)
Gene Aliases
1110017O22Rik, Ccdc72
Chromosome Location
Chr. 9: 109077988 - 109082381 on Build GRCm38
UniGene ID
Mm.2395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183250.2 NP_899073.1 2, 3 109
GenBank mRNA AK002863.1 - 2, 3 110
AK003762.1 - 2, 3 110
AK160705.1 - 2, 3 107
AK169860.1 - 2, 3 103
BC034602.1 - 2, 3 91
BC054363.1 - 2, 3 82
BC090400.1 - 2, 3 82
BC106104.1 - 2, 3 81 5029265
BC116987.1 - 2, 3 105 40126306
BC116989.1 - 2, 3 105 40126308

View Full Product Details