Gene Transcripts

Gene Symbol
Asf1a
Entrez Gene ID
66403
Gene Name
anti-silencing function 1A histone chaperone
Gene Aliases
2310079C17Rik, C79052
Chromosome Location
Chr. 10: 53596961 - 53609225 on Build GRCm38
UniGene ID
Mm.272989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025541.3 NP_079817.1 3 534
GenBank mRNA AK007804.1 - 3 390
AK010210.1 - 3 544
AK012376.1 - 3 525
AK031513.1 - 3 500
AK088618.1 - 3 477
BC027628.1 - 3 326 1178866

View Full Product Details