Gene Transcripts

Gene Symbol
Zmat2
Entrez Gene ID
66492
Gene Name
zinc finger, matrin type 2
Gene Aliases
2610510D14Rik, 2900082I05Rik
Chromosome Location
Chr. 18: 36793923 - 36799660 on Build GRCm38
UniGene ID
Mm.251155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025594.3 NP_079870.1 3, 4 248
GenBank mRNA AK008717.1 - 3, 4 241
AK012091.1 - 3, 4 244
BC089501.1 - 3, 4 243 6594761

View Full Product Details