Gene Transcripts

Gene Symbol
Smarca2
Entrez Gene ID
67155
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2
Gene Aliases
2610209L14Rik, SNF2alpha, Snf2l2, brahma, brm
Chromosome Location
Chr. 19: 26605089 - 26778321 on Build GRCm38
UniGene ID
Mm.491146
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011416.2 NP_035546.2 30 4513
NM_026003.2 NP_080279.1 6 662
GenBank mRNA AK011935.1 - 6 607
AK147322.1 - 30 4662
BC004653.1 - 2 160
BC022975.1 - 8 1139
BC056199.1 - 20 2611
BC075641.1 - 30 4495 30544092

View Full Product Details