Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144517.4 | NP_653100.1 | 6 | 548 | |
GenBank mRNA | AK012836.1 | - | 6 | 500 | |
AK076128.1 | - | 6 | 496 | ||
AK083084.1 | - | 6 | 519 | ||
AK177065.1 | - | 6 | 435 | ||
BC020138.1 | - | 6 | 507 | 3983701 |