Gene Transcripts

Gene Symbol
Myl12a
Entrez Gene ID
67268
Gene Name
myosin, light chain 12A, regulatory, non-sarcomeric
Gene Aliases
2900073G15Rik, AI838292, C80200
Chromosome Location
Chr. 17: 70993793 - 71002533 on Build GRCm38
UniGene ID
Mm.489605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026064.2 NP_080340.2 4 1738
GenBank mRNA AK013776.1 - 4 1425
AK018348.1 - 4 1393
BC010265.1 - 3 1323 3588394

View Full Product Details