Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026176.3 | NP_080452.2 | 4 | 985 | |
GenBank mRNA | AK004704.1 | - | 4 | 1698 | |
AK028668.1 | - | 4 | 875 | ||
AK028814.1 | - | 4 | 875 | ||
AK133747.1 | - | 4 | 895 | ||
AK166983.1 | - | 4 | 875 | ||
BC006578.1 | - | 4 | 885 | 3586612 |