Gene Transcripts

Gene Symbol
Spert
Entrez Gene ID
67926
Gene Name
spermatid associated
Gene Aliases
1700086N05Rik, AI596460, Nurit
Chromosome Location
Chr. 14: 75582834 - 75593177 on Build GRCm38
UniGene ID
Mm.23361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164139.1 NP_001157611.1 3 920
NM_001164140.1 NP_001157612.1 2 839
NM_001164141.1 NP_001157613.1 3 928
NM_026457.2 NP_080733.2 2 778
GenBank mRNA AK006329.1 - 1 508
AK007022.1 - 2 757
AY092416.1 - 2 912
BC109147.2 - 2 757 40045644

View Full Product Details