Gene Transcripts
- Gene Symbol
- Otub2
- Entrez Gene ID
- 68149
- Gene Name
- OTU domain, ubiquitin aldehyde binding 2
- Gene Aliases
- 2010015L18Rik, 4930586I02Rik, AI413508, AW557219, OTB2, OTU2
- Chromosome Location
- Chr. 12: 103376894 - 103406351 on Build GRCm38
- UniGene ID
- Mm.127378
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177841.1 |
NP_001171312.1
|
5, 6 |
911 |
|
NM_026580.4 |
NP_080856.1
|
4, 5 |
485 |
|
GenBank mRNA |
AK006054.1 |
- |
4, 5 |
407 |
|
AK006346.1 |
- |
4, 5 |
437 |
|
AK006469.1 |
- |
4, 5 |
434 |
|
AK008258.2 |
- |
4, 5 |
369 |
|
AK016449.1 |
- |
4, 5 |
469 |
|
AK019830.1 |
- |
4, 5 |
462 |
|
AK076336.1 |
- |
5, 6 |
915 |
|
AK158758.1 |
- |
4, 5 |
768 |
|
AK160505.1 |
- |
4, 5 |
464 |
|
BC049551.1 |
- |
4, 5 |
468 |
6771956 |
View Full Product Details