Gene Transcripts

Gene Symbol
Ggct
Entrez Gene ID
110175
Gene Name
gamma-glutamyl cyclotransferase
Gene Aliases
A030007L17Rik, AA673177, Gctg, Ggc
Chromosome Location
Chr. 6: 54985095 - 54992965 on Build GRCm38
UniGene ID
Mm.294708
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026637.3 NP_080913.1 4 548
GenBank mRNA AK008719.1 - 4 553
AK028830.1 - 550
AK050049.1 - 4 633
AK079465.1 - 4 550
AK141474.1 - 598
BC080818.1 - 4 528 30286931

View Full Product Details