Gene Transcripts

Gene Symbol
Ccdc88c
Entrez Gene ID
68339
Gene Name
coiled-coil domain containing 88C
Gene Aliases
0610010D24Rik, AV000935, AW324073, Daple, mKIAA1509
Chromosome Location
Chr. 12: 100911521 - 101028983 on Build GRCm38
UniGene ID
Mm.156217
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026681.4 NP_080957.2 26, 27 4756
GenBank mRNA AK173188.1 - 13, 14 2570
AY339881.1 - 26, 27 4757

View Full Product Details