Gene Transcripts

Gene Symbol
Fam96b
Entrez Gene ID
68523
Gene Name
family with sequence similarity 96, member B
Gene Aliases
1110019N10Rik
Chromosome Location
Chr. 8: 104639839 - 104641728 on Build GRCm38
UniGene ID
Mm.279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026753.2 NP_081029.1 1, 2 176
GenBank mRNA AK003830.1 - 1, 2 179
BC055880.1 - 1, 2 150 4190754

View Full Product Details