Gene Transcripts

Gene Symbol
Ctdspl
Entrez Gene ID
69274
Gene Name
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like
Gene Aliases
2810418J22Rik, AI426263, HYA22, SCP3
Chromosome Location
Chr. 9: 118926536 - 119044119 on Build GRCm38
UniGene ID
Mm.439928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133710.3 NP_598471.3 8 1482
GenBank mRNA AJ344340.1 - 8 1564
AK138484.1 - 1 212
AK171161.1 - 1 212

View Full Product Details