Gene Transcripts

Gene Symbol
H2bfm
Entrez Gene ID
69389
Gene Name
H2B histone family, member M
Gene Aliases
1700014N06Rik, H2bfw, H2bl2
Chromosome Location
Chr. X: 136927325 - 136928373 on Build GRCm38
UniGene ID
Mm.302721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027067.2 NP_081343.1 1 449
GenBank mRNA AK005982.1 - 1 450
BC117076.1 - 1 440 40126395
BC117080.1 - 1 440 40126399

View Full Product Details