Gene Transcripts

Gene Symbol
Bst2
Entrez Gene ID
69550
Gene Name
bone marrow stromal cell antigen 2
Gene Aliases
2310015I10Rik, Bst-2, C87040, CD317, DAMP-1, GREG
Chromosome Location
Chr. 8: 71534259 - 71537437 on Build GRCm38
UniGene ID
Mm.260325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198095.2 NP_932763.1 4 441
GenBank mRNA AK009361.1 - 4 241
AK154399.1 - 4 434
BC027328.1 - 4 475 4009434
BC056638.1 - 4 448 5065707
BC087949.1 - 4 433 6768735

View Full Product Details