Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243584.1 | NP_001230513.1 | 4 | 541 | |
NM_001243586.1 | NP_001230515.1 | 3 | 825 | ||
NM_001243587.1 | NP_001230516.1 | 4 | 541 | ||
NM_027162.4 | NP_081438.4 | 3 | 825 | ||
GenBank mRNA | AK003708.1 | - | 602 | ||
AK010175.1 | - | 4 | 495 | ||
AK150749.1 | - | 4 | 495 | ||
BC019515.1 | - | 4 | 452 | 4460171 | |
BC026740.1 | - | 4 | 501 | 4015105 | |
BC055812.1 | - | 3 | 825 | 4934728 | |
CX210298.1 | - | 4 | 525 |