Gene Transcripts

Gene Symbol
Nup37
Entrez Gene ID
69736
Gene Name
nucleoporin 37
Gene Aliases
2410003L22Rik, 2810039M17Rik
Chromosome Location
Chr. 10: 88146976 - 88178395 on Build GRCm38
UniGene ID
Mm.222184
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027191.2 NP_081467.2 7 856
NM_028334.4 NP_082610.1 7 874
GenBank mRNA AK010370.1 - 7 800
AK012890.1 - 7 870
AK040463.1 - 7 857
AK165537.1 - 7 820
BC011102.1 - 7 840 3982076
BC032180.1 - 6 784 5151847

View Full Product Details