Gene Transcripts
- Gene Symbol
- Sh2d6
- Entrez Gene ID
- 71130
- Gene Name
- SH2 domain containing 6
- Gene Aliases
- Chromosome Location
- Chr. 6: 72513652 - 72534207 on Build GRCm38
- UniGene ID
- Mm.128597
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016886.1 |
- |
2, 3 |
296 |
|
BC100525.1 |
- |
2, 3 |
253 |
30920200 |
View Full Product Details