Gene Transcripts

Gene Symbol
Fam13c
Entrez Gene ID
71721
Gene Name
family with sequence similarity 13, member C
Gene Aliases
1200015N20Rik, C030038O19Rik, mKIAA1796
Chromosome Location
Chr. 10: 70440668 - 70558732 on Build GRCm38
UniGene ID
Mm.477813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143776.1 NP_001137248.1 7 887
NM_001143777.1 NP_001137249.1 8 991
NM_024244.4 NP_077206.3 7 887
GenBank mRNA AK004797.1 - 7 871
AK021133.1 - 8 830
AK032053.1 - 8 803
AK036299.1 - 8 954
AK041147.1 - 7 888
AK044049.1 - 8 914
AK082202.1 - 7 883
AK220421.1 - 8 827
BC129839.1 - 7 843 40060810
BC129840.1 - 7 843 40060811

View Full Product Details