Gene Transcripts

Gene Symbol
Cpn2
Entrez Gene ID
71756
Gene Name
carboxypeptidase N, polypeptide 2
Gene Aliases
1300018K11Rik
Chromosome Location
Chr. 16: 30256378 - 30267614 on Build GRCm38
UniGene ID
Mm.290614
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027904.3 NP_082180.2 2 876
GenBank mRNA AK005049.1 - 2 878
BC025836.1 - 2 843
BC081550.1 - 1 395
BC138287.1 - 2 963 8861306
BC138288.1 - 2 963 8861307

View Full Product Details