Gene Transcripts

Gene Symbol
March8
Entrez Gene ID
71779
Gene Name
membrane-associated ring finger (C3HC4) 8
Gene Aliases
1300017E09Rik, MARCH-VIII, Mir
Chromosome Location
Chr. 6: 116337884 - 116409540 on Build GRCm38
UniGene ID
Mm.27064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK005032.2 - 7 894
AK154976.1 - 8 1086
AK208723.1 - 2 236
BC050908.1 - 8 1069 6416955
BC053090.1 - 7 1154 5709246

View Full Product Details