Gene Transcripts
- Gene Symbol
- Snx10
- Entrez Gene ID
- 71982
- Gene Name
- sorting nexin 10
- Gene Aliases
- 2410004M09Rik
- Chromosome Location
- Chr. 6: 51523902 - 51590679 on Build GRCm38
- UniGene ID
- Mm.294166
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127348.1 |
NP_001120820.1
|
4, 5 |
358 |
|
NM_001127349.1 |
NP_001120821.1
|
4, 5 |
363 |
|
NM_028035.4 |
NP_082311.3
|
4, 5 |
461 |
|
GenBank mRNA |
AK010399.1 |
- |
4, 5 |
306 |
|
AK028163.1 |
- |
4, 5 |
462 |
|
AK150704.1 |
- |
4, 5 |
366 |
|
AK150878.1 |
- |
4, 5 |
280 |
|
AK150999.1 |
- |
4, 5 |
366 |
|
AK151320.1 |
- |
4, 5 |
366 |
|
AK151520.1 |
- |
4, 5 |
366 |
|
AK152004.1 |
- |
4, 5 |
284 |
|
AK152825.1 |
- |
4, 5 |
338 |
|
AK152949.1 |
- |
4, 5 |
354 |
|
AK153127.1 |
- |
4, 5 |
366 |
|
AK153356.1 |
- |
4, 5 |
316 |
|
AK166121.1 |
- |
4, 5 |
331 |
|
AK169110.1 |
- |
4, 5 |
297 |
|
BC010334.1 |
- |
4, 5 |
295 |
2655824 |
CT010276.1 |
- |
3, 4 |
208 |
|
View Full Product Details