Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028097.3 | NP_082373.1 | 7 | 964 | |
GenBank mRNA | AK008493.1 | - | 7 | 998 | |
AK048210.1 | - | 7 | 964 | ||
AK084881.1 | - | 7 | 998 | ||
AK088600.1 | - | 8 | 1134 | ||
AK147765.1 | - | 7 | 997 | ||
AK162563.1 | - | 7 | 997 | ||
AK165916.1 | - | 7 | 1026 | ||
AK166237.1 | - | 7 | 1050 | ||
BC016240.1 | - | 7 | 982 | 4506688 |