Gene Transcripts
- Gene Symbol
- MUC4
- Entrez Gene ID
- 4585
- Gene Name
- mucin 4, cell surface associated
- Gene Aliases
- ASGP, HSA276359, MUC-4
- Chromosome Location
- Chr. 3: 195746765 - 195811973 on Build GRCh38
- UniGene ID
- Hs.369646
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001322468.1 |
NP_001309397.1
|
|
21690 |
|
NM_004532.5 |
NP_004523.3
|
19, 20 |
2964 |
|
NM_018406.6 |
NP_060876.5
|
|
15672 |
|
NM_138297.4 |
NP_612154.2
|
18, 19 |
2811 |
|
GenBank mRNA |
AJ010901.1 |
- |
19, 20 |
2748 |
|
AJ242541.1 |
- |
19, 20 |
2813 |
|
AJ242542.1 |
- |
18, 19 |
2673 |
|
AJ242543.1 |
- |
18, 19 |
2512 |
|
AJ242544.1 |
- |
19, 20 |
2644 |
|
AJ242545.1 |
- |
18, 19 |
2583 |
|
AJ242546.1 |
- |
16, 17 |
2157 |
|
AJ242547.1 |
- |
18, 19 |
2588 |
|
AJ242548.1 |
- |
15, 16 |
2070 |
|
AJ242549.1 |
- |
19, 20 |
2848 |
|
AJ242550.1 |
- |
18, 19 |
2695 |
|
AJ276359.1 |
- |
21, 22 |
5858 |
|
AJ277412.1 |
- |
20, 21 |
5702 |
|
AJ277505.1 |
- |
19, 20 |
5599 |
|
AJ400633.1 |
- |
21, 22 |
5872 |
|
AJ400849.1 |
- |
21, 22 |
5782 |
|
AJ400850.1 |
- |
21, 22 |
5782 |
|
AJ400851.1 |
- |
19, 20 |
5498 |
|
AJ400852.1 |
- |
20, 21 |
5626 |
|
AJ400853.1 |
- |
20, 21 |
5598 |
|
AJ400854.1 |
- |
20, 21 |
5703 |
|
AJ400855.1 |
- |
20, 21 |
5674 |
|
AJ400856.1 |
- |
21, 22 |
5829 |
|
AJ400857.1 |
- |
20, 21 |
5698 |
|
AJ400858.1 |
- |
21, 22 |
5852 |
|
View Full Product Details