Gene Transcripts

Gene Symbol
MVK
Entrez Gene ID
4598
Gene Name
mevalonate kinase
Gene Aliases
LRBP, MK, MVLK, POROK3
Chromosome Location
Chr. 12: 109573285 - 109597270 on Build GRCh38
UniGene ID
Hs.130607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000431.3 NP_000422.1 3, 4 429
NM_001114185.2 NP_001107657.1 3, 4 420
NM_001301182.1 NP_001288111.1 3, 4 429
XM_011538372.2 XP_011536674.2 3, 4 537
XM_017019313.1 XP_016874802.1 3, 4 382
XM_017019314.1 XP_016874803.1 3, 4 895
GenBank mRNA AB209722.1 - 3, 4 312
AF217536.1 - 3, 4 343
AK295338.1 - 3, 4 287
AK315678.1 - 3, 4 308
BC016140.1 - 3, 4 272 3921543
DC421549.1 - 3, 4 429
KU178205.1 - 2, 3 236
KU178206.1 - 2, 3 236
M88468.1 - 3, 4 327

View Full Product Details