Gene Transcripts

Gene Symbol
Tbcc
Entrez Gene ID
72726
Gene Name
tubulin-specific chaperone C
Gene Aliases
2810055C19Rik
Chromosome Location
Chr. 17: 46890621 - 46892463 on Build GRCm38
UniGene ID
Mm.33780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178385.3 NP_848472.2 1 980
GenBank mRNA AK012950.1 - 1 925
AK132036.1 - 930
BC019476.1 - 1 918 4240883
BC083164.1 - 1 912 5390600

View Full Product Details