Gene Transcripts

Gene Symbol
Zdhhc4
Entrez Gene ID
72881
Gene Name
zinc finger, DHHC domain containing 4
Gene Aliases
1810021D01Rik, 2900029I10Rik
Chromosome Location
Chr. 5: 143316489 - 143329252 on Build GRCm38
UniGene ID
Mm.261606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028379.5 NP_082655.1 6 725
GenBank mRNA AK007572.1 - 4 711
AK013609.1 - 6 712
AK156167.1 - 6 708
BC019523.1 - 6 707 4163453

View Full Product Details