Gene Transcripts

Gene Symbol
Rbm19
Entrez Gene ID
74111
Gene Name
RNA binding motif protein 19
Gene Aliases
1200009A02Rik, AV302714, NPO
Chromosome Location
Chr. 5: 120116513 - 120198971 on Build GRCm38
UniGene ID
Mm.41022
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028762.1 NP_083038.1 16, 17 2154
GenBank mRNA AK004657.1 - 7, 8 969
AK146493.1 - 16, 17 2187
AK167100.1 - 16, 17 2151
BC025619.1 - 16, 17 2154 5320947
BC034010.1 - 16, 17 2120 4225239

View Full Product Details