Gene Transcripts

Gene Symbol
Tspan17
Entrez Gene ID
74257
Gene Name
tetraspanin 17
Gene Aliases
2210021G21Rik, AI047581, Fbxo23, Tm4sf17
Chromosome Location
Chr. 13: 54789352 - 54796896 on Build GRCm38
UniGene ID
Mm.32883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028841.3 NP_083117.2 4 608
GenBank mRNA AI019499.1 - 1 79
AK008761.1 - 4 553
AK166272.1 - 4 611
AK171691.1 - 4 557
AK219107.1 - 1 46
BC010346.1 - 4 553 2811935
BY734818.1 - 4 607

View Full Product Details