Gene Transcripts

Gene Symbol
Pck2
Entrez Gene ID
74551
Gene Name
phosphoenolpyruvate carboxykinase 2 (mitochondrial)
Gene Aliases
1810010O14Rik, 9130022B02Rik, PEPCK-M
Chromosome Location
Chr. 14: 55540263 - 55550777 on Build GRCm38
UniGene ID
Mm.29856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028994.2 NP_083270.1 10 1942
GenBank mRNA AK007429.1 - 7 1134
AK030501.1 - 10 1943
AK031612.1 - 10 1787
AK034927.1 - 10 1821
AK142561.1 - 4 694
AK147972.1 - 10 1796
AK165094.1 - 10 1820
AK170242.1 - 10 1796
AK199613.1 - 1 100
BC010318.1 - 10 1805 3596227
BC011126.1 - 9 2104
BC023437.1 - 8 1410
BC028326.1 - 5 743

View Full Product Details