Gene Transcripts

Gene Symbol
Sf3a3
Entrez Gene ID
75062
Gene Name
splicing factor 3a, subunit 3
Gene Aliases
4930512K19Rik, 60kDa
Chromosome Location
Chr. 4: 124714861 - 124732423 on Build GRCm38
UniGene ID
Mm.25779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029157.3 NP_083433.1 10 827
GenBank mRNA AK030743.1 - 10 862
AK150733.1 - 10 825
AK151141.1 - 10 825
AK151382.1 - 10 825
AK161646.1 - 10 848
AK168966.1 - 10 851
AK169324.1 - 10 871
BC009141.1 - 10 827 3598647
BC092058.1 - 10 859 4017388

View Full Product Details