Gene Transcripts

Gene Symbol
Ccl27a
Entrez Gene ID
20301
Gene Name
chemokine (C-C motif) ligand 27A
Gene Aliases
ALP, AW558992, CTACK, CTAK, Ccl27, ESkine, ILC, PESKY, Scya27, Scya27a
Chromosome Location
Chr. 4: 41769467 - 41774210 on Build GRCm38
UniGene ID
Mm.335946
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164044.1 NP_001157516.1 4 626
NM_001164045.1 NP_001157517.1 3 847

View Full Product Details