Gene Transcripts

Gene Symbol
Fastkd2
Entrez Gene ID
75619
Gene Name
FAST kinase domains 2
Gene Aliases
2810421I24Rik
Chromosome Location
Chr. 1: 63730599 - 63753385 on Build GRCm38
UniGene ID
Mm.30965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172422.3 NP_766010.1 3 968
GenBank mRNA AK035322.1 - 3 969
AK038293.1 - 3 952
AK084971.1 - 3 943
AK087156.1 - 3 950
AK129251.1 - 2 906
AK148224.1 - 952
AK153916.1 - 2 911
AK166378.1 - 3 953
AK167714.1 - 3 950
BC008271.1 - 2 909 3603187
BC057208.1 - 2 902 5328766
BC080813.1 - 2 908 6394260

View Full Product Details