Gene Transcripts

Gene Symbol
Cldn22
Entrez Gene ID
75677
Gene Name
claudin 22
Gene Aliases
2210404A22Rik, 5330431C14Rik, 9530051B05Rik
Chromosome Location
Chr. 8: 47824482 - 47825475 on Build GRCm38
UniGene ID
Mm.390033
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029383.1 NP_083659.1 1 153
GenBank mRNA AK008821.1 - 1 154

Gene Symbol
Cldn24
Entrez Gene ID
100039801
Gene Name
claudin 24
Gene Aliases
ENSMUSG00000061974, Gm10107
Chromosome Location
Chr. 8: 47822143 - 47822805 on Build GRCm38
UniGene ID
Mm.471595
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111318.1 NP_001104788.1 1 106

View Full Product Details