Gene Transcripts

Gene Symbol
NEK1
Entrez Gene ID
4750
Gene Name
NIMA related kinase 1
Gene Aliases
NY-REN-55, SRPS2, SRPS2A, SRTD6
Chromosome Location
Chr. 4: 169393270 - 169612629 on Build GRCh38
UniGene ID
Hs.481181
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199397.1 NP_001186326.1 32 3874
NM_001199398.1 NP_001186327.1 31 3742
NM_001199399.1 NP_001186328.1 29 3583
NM_001199400.1 NP_001186329.1 29 3736
NM_012224.2 NP_036356.1 30 3868
XM_006714228.1 XP_006714291.1 31 3699
XM_011532003.1 XP_011530305.1 31 3792
XM_011532004.1 XP_011530306.1 30 3660
XM_017008249.1 XP_016863738.1 31 3778
XM_017008250.1 XP_016863739.1 30 3896
XM_017008251.1 XP_016863740.1 30 3694
XM_017008252.1 XP_016863741.1 28 3714
XM_017008253.1 XP_016863742.1 31 3724
XM_017008254.1 XP_016863743.1 24 2834
GenBank mRNA AB067488.1 - 30 3704
AF155113.1 - 14 1752
AK025658.1 - 5 622
AL050385.1 - 31 3698
BC037790.2 - 4 534
BC114491.1 - 29 3343 40082305
CR933642.1 - 32 3832
CR936672.1 - 29 3694

View Full Product Details